Order Entry
Ireland
ContactUsLinkComponent
Comfort rack for chest freezers for 14 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 592x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24442B
Supplier:  TENAK
New Product
Comfort rack for chest freezers for 14 cryoboxes, 40mm high, stainless steel, with spring lock, dimensions (HxDxW) 592x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE24442B
Supplier:  TENAK