Standard rack for upright freezers for (HxD) 4x5=20 cryoboxes, 75mm high, stainless steel, dimensions (HxDxW) 319x683x139mm, deliver ed including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE23194B10
Supplier: TENAK