Order Entry
Ireland
ContactUsLinkComponent
Standard rack for upright freezers for (HxD) 7x5=35 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 253x683x139mm, deliver ed including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE23084B10
 :  TENAK
New Product
Standard rack for upright freezers for (HxD) 7x5=35 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 253x683x139mm, deliver ed including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
  TENATE23084B10
 :  TENAK