Order Entry
Ireland
ContactUsLinkComponent
Standard rack for upright freezers for (HxD) 4x4=16 cryoboxes, 40mm high, stainless steel, dimensions (HxDxW) 171x561x139mm, deliver ed including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE23022B10
Supplier:  TENAK
New Product
Standard rack for upright freezers for (HxD) 4x4=16 cryoboxes, 40mm high, stainless steel, dimensions (HxDxW) 171x561x139mm, deliver ed including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE23022B10
Supplier:  TENAK