Order Entry
Ireland
ContactUsLinkComponent
Side up rack for both chest and upright freezers for 3 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 140x420x36mm, deliv ered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE21390B
Supplier:  TENAK
New Product
Side up rack for both chest and upright freezers for 3 cryoboxes, 32mm high, stainless steel, dimensions (HxDxW) 140x420x36mm, deliv ered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE21390B
Supplier:  TENAK