Order Entry
Ireland
ContactUsLinkComponent
Classic rack for chest freezers for 11 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 466x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE21380B
Supplier:  TENAK
New Product
Classic rack for chest freezers for 11 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 466x140x140mm, de livered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE21380B
Supplier:  TENAK