Order Entry
Ireland
ContactUsLinkComponent
Classic rack for chest freezers for 11 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 466x140x140mm 1 * 1 items
Catalog # TENATE21380
Supplier:  TENAK
New Product
Classic rack for chest freezers for 11 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 466x140x140mm 1 * 1 items
Catalog # TENATE21380
Supplier:  TENAK