Order Entry
Ireland
ContactUsLinkComponent
Classic rack for chest freezers for 9 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 382x140x140mm, del ivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE21376B10
Supplier:  TENAK
New Product
Classic rack for chest freezers for 9 cryoboxes, 40mm high, stainless steel, with locking rod, dimensions (HxDxW) 382x140x140mm, del ivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE21376B10
Supplier:  TENAK