Order Entry
Ireland
ContactUsLinkComponent
Classic rack for chest freezers for 20 cryoboxes, 32mm high, stainless steel, with locking rod, dimensions (HxDxW) 714x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE21362B10
Supplier:  TENAK
New Product
Classic rack for chest freezers for 20 cryoboxes, 32mm high, stainless steel, with locking rod, dimensions (HxDxW) 714x140x140mm, de livered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE21362B10
Supplier:  TENAK