Order Entry
Ireland
ContactUsLinkComponent
Classic rack for chest freezers for 8 cryoboxes, 32mm high, stainless steel, with locking rod, dimensions (HxDxW) 288x140x140mm, del ivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE21350B
Supplier:  TENAK
New Product
Classic rack for chest freezers for 8 cryoboxes, 32mm high, stainless steel, with locking rod, dimensions (HxDxW) 288x140x140mm, del ivered including standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE21350B
Supplier:  TENAK