Order Entry
Ireland
ContactUsLinkComponent
Eco Alu MTP rack for upright freezers for SBS format (HxD) 2x5=10 plates, 90mm high, aluminium, dimensions (HxDxW) 185x450x135mm 1 * 1 items
Catalog # TENATE15666
Supplier:  TENAK
New Product
Eco Alu MTP rack for upright freezers for SBS format (HxD) 2x5=10 plates, 90mm high, aluminium, dimensions (HxDxW) 185x450x135mm 1 * 1 items
Catalog # TENATE15666
Supplier:  TENAK