Order Entry
Ireland
ContactUsLinkComponent
Eco Alu MTP rack for upright freezers for SBS format (HxD) 3x6=18 plates, 55mm high, aluminium, dimensions (HxDxW) 172x540x135mm 1 * 1 items
Catalog # TENATE15638
Supplier:  TENAK
New Product
Eco Alu MTP rack for upright freezers for SBS format (HxD) 3x6=18 plates, 55mm high, aluminium, dimensions (HxDxW) 172x540x135mm 1 * 1 items
Catalog # TENATE15638
Supplier:  TENAK