Order Entry
Ireland
ContactUsLinkComponent
Eco Alu MTP rack for upright freezers for SBS format (HxD) 4x5=20 plates, 55mm high, aluminium, dimensions (HxDxW) 229x450x135mm 1 * 1 items
Catalog # TENATE15630
Supplier:  TENAK
New Product
Eco Alu MTP rack for upright freezers for SBS format (HxD) 4x5=20 plates, 55mm high, aluminium, dimensions (HxDxW) 229x450x135mm 1 * 1 items
Catalog # TENATE15630
Supplier:  TENAK