Order Entry
Ireland
ContactUsLinkComponent
Eco Alu MTP rack for upright freezers for SBS format (HxD) 6x5=30 plates, 35mm high, aluminium, dimensions (HxDxW) 223x450x135mm 1 * 1 items
Catalog # TENATE15312
Supplier:  TENAK
New Product
Eco Alu MTP rack for upright freezers for SBS format (HxD) 6x5=30 plates, 35mm high, aluminium, dimensions (HxDxW) 223x450x135mm 1 * 1 items
Catalog # TENATE15312
Supplier:  TENAK