Order Entry
Ireland
ContactUsLinkComponent
Eco AluRack for chest freezers for 5 cryoboxes, 75mm high, aluminium, dimensions (HxDxW) 406x140x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11536B10
Supplier:  TENAK
New Product
Eco AluRack for chest freezers for 5 cryoboxes, 75mm high, aluminium, dimensions (HxDxW) 406x140x140mm, delivered including standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11536B10
Supplier:  TENAK