Order Entry
Ireland
ContactUsLinkComponent
Eco AluRack for upright freezers for (HxD) 3x2=6 cryoboxes, 75mm high, aluminium, dimensions (HxDxW) 240x280x139mm, delivered includ ing standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11260B10
Supplier:  TENAK
New Product
Eco AluRack for upright freezers for (HxD) 3x2=6 cryoboxes, 75mm high, aluminium, dimensions (HxDxW) 240x280x139mm, delivered includ ing standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11260B10
Supplier:  TENAK