Order Entry
Ireland
ContactUsLinkComponent
Eco AluRack for upright freezers for (HxD) 7x3=21 cryoboxes, 32mm high, aluminium, dimensions (HxDxW) 253x425x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11124B10
Supplier:  TENAK
New Product
Eco AluRack for upright freezers for (HxD) 7x3=21 cryoboxes, 32mm high, aluminium, dimensions (HxDxW) 253x425x139mm, delivered inclu ding standard white cardboard boxes and 10x10 cell dividers 1 * 1 items
Catalog # TENATE11124B10
Supplier:  TENAK