Order Entry
Ireland
ContactUsLinkComponent
Eco AluRack for upright freezers for (HxD) 9x5=45 cryoboxes, 40mm high, aluminium, dimensions (HxDxW) 384x683x139mm, delivered inclu ding standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE11044B
Supplier:  TENAK
New Product
Eco AluRack for upright freezers for (HxD) 9x5=45 cryoboxes, 40mm high, aluminium, dimensions (HxDxW) 384x683x139mm, delivered inclu ding standard white cardboard boxes and 9x9 cell dividers 1 * 1 items
Catalog # TENATE11044B
Supplier:  TENAK