GromSil 120 ODS-3 CP, 10um, 10x20mm, Fujiplate guard column 1 * 1 items
Catalog # MAISGSOD31012V0120
Supplier: DR MAISCH
New Product
GromSil 120 ODS-3 CP, 10um, 10x20mm, Fujiplate guard column 1 * 1 items
Catalog # MAISGSOD31012V0120
Supplier: DR MAISCH
New Product
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Transfer pipettes
Supplier: RATIOLAB
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
DURAN® GLS 80® Production and Storage Bottles
Supplier: DWK Life Sciences