PILZ HEIZHAUBE CLASSIC LINE SERIE G2H 1 * 1 items
Catalog # WINKW0589-1454
Supplier: WINKLER
New Product
PILZ HEIZHAUBE CLASSIC LINE SERIE G2H 1 * 1 items
Catalog # WINKW0589-1454
Supplier: WINKLER
New Product
Frequently Bought Together
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
VWR® Reagent Reservoirs
Supplier: VWR Collection
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Nitric acid 0.2 mol/l (0.2 N)
Supplier: VWR Collection