Order Entry
Ireland
ContactUsLinkComponent
Muffle furnace lt5/14/c550 tmax1400 with with lift door and protective gas connection, over-temperature limiter and thermocouple hol e in furnace door 1 * 1 items
Catalog # NABEL-055H4ANOGTY
Supplier:  NABERTHERM
New Product
Muffle furnace lt5/14/c550 tmax1400 with with lift door and protective gas connection, over-temperature limiter and thermocouple hol e in furnace door 1 * 1 items
Catalog # NABEL-055H4ANOGTY
Supplier:  NABERTHERM