2.5LT METHANOL BAKER ANALYZED LC/MS 1 * 2,5 L
Catalog # MSPP15514065
Supplier: MARKET SOURCE PART PROCESS
2.5LT METHANOL BAKER ANALYZED LC/MS 1 * 2,5 L
Catalog # MSPP15514065
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Autoclavable Bags, Polypropylene, Clear
Supplier: VWR Collection
VWR® Autoclavable Biohazard Bags, Polypropylene, Orange
Supplier: VWR Collection
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
JERRYCAN 10 L HDPE NATURAL W SCREWCAP 1 * 10 items
Supplier: RIXIUS
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals