Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ROVERS ORCELLEX BRUSH RT STERILE Remover Tube is a feature on the handle to easily release the brush head without contamination 1 * 100 items
Catalog # ROVE380380331
Supplier:
Rovers
New Product
Expand image
ROVERS ORCELLEX BRUSH RT STERILE Remover Tube is a feature on the handle to easily release the brush head without contamination 1 * 100 items
Catalog # ROVE380380331
Supplier:
Rovers
New Product
Frequently Bought Together
REAGECON R1 COLOUR REFERENCE SOLUTION ACCORDING TO EUROPEAN PHARMACOPOEIA (EP) 1 * 100 mL
Supplier: MARKET SOURCE PART PROCESS
Trichloroethylene Analytical standard, Fluka™
Supplier: Honeywell Chemicals