Fixative,Bouins Fluid,500ML 1 * 1 items
Catalog # SHAN88038
Supplier: Epredia
New Product
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Pyridine ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.9%, TraceSELECT™ for inorganic trace analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Aniline ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals