Click's Medium is a modification of Eagle's Essential Medium with Hanks' Salts (HMEM) and contains higher concentrations of essentia l amino acids, in addition to non-essential amino acids, sodium pyruvate, and nucleic acid precursors. 1 * 500 mL
Catalog # SHBT9195
Supplier: Shenandoah Biotechnology