PRIME-XV FreezIS DMSO-Free solution is recommended mainly for the cryopreservation of human mesenchymal stromal cells (MSCs), CD34+ hematopoietic stem cells (HSCs), T cells derived from peripheral blood mononuclear cells (PBMCs), and potentially othe 1 * 100 mL
Catalog # SHBT91140-100ML
Supplier: Shenandoah Biotechnology