PRIME-XV Neural Basal Medium is a chemically-defined, animal component- free medium that supports a range of applications for neural stem cells. Designed and manufactured to facilitate transfer from research to clinic:FDA, Federal, and State register 1 * 500 mL
Catalog # SHBT91201
Supplier: Shenandoah Biotechnology