Order Entry
Ireland
ContactUsLinkComponent
,,Multi-element calibration standard-4B - 12 components: 10ug/ml each of B Ge Mo Nb P Re S Si Ta Ti W Zr in HNO3 2%/ tr. HF ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 36 months.,, ,,Ambient Temp.,, 1 * 500 mL
Catalog # CMPLRM004671L5
New Product
,,Multi-element calibration standard-4B - 12 components: 10ug/ml each of B Ge Mo Nb P Re S Si Ta Ti W Zr in HNO3 2%/ tr. HF ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 36 months.,, ,,Ambient Temp.,, 1 * 500 mL
Catalog # CMPLRM004671L5