Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CORONAVIRUS (SARS-COV NP) ANTIGEN (AA 1-49, 192-220), (RECOMBINANT) 1 * 100 µG
Catalog # ENZOENZPRT2940100
Supplier:
ENZO LIFE SCIENCES
Expand image
CORONAVIRUS (SARS-COV NP) ANTIGEN (AA 1-49, 192-220), (RECOMBINANT) 1 * 100 µG
Catalog # ENZOENZPRT2940100
Supplier:
ENZO LIFE SCIENCES
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Petri dishes
Supplier: Greiner Bio-One
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF