RUCAPARIB PHOSPHATE 1 * 100 mg
Catalog # ENZOENZCHM2360100
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
Ammonium iron(II) sulphate hexahydrate ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium salicylate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium sulphate, anhydrous 98.5-101.0%, fine powder USP, Ph. Eur.
Supplier: VWR Chemicals
Potassium thiocyanate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Lithium bromide ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium perchlorate monohydrate 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Lithium chloride ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium thiosulphate, anhydrous ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals