AMPIVIEW DIG (ANTI-RABBIT) DETECTION AP-RED KIT 1 * 20 Assays
Catalog # ENZOENZACC1600020
Supplier: ENZO LIFE SCIENCES
AMPIVIEW DIG (ANTI-RABBIT) DETECTION AP-RED KIT 1 * 20 Assays
Catalog # ENZOENZACC1600020
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
96-Well clear polystyrene microplates, Corning®
Supplier: Corning
VWR®, Microcentrifuge Tubes with Flat Screw Caps
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
Sterile Serological Pipettes
Supplier: EPPENDORF
Cell culture flasks, Falcon®
Supplier: Corning
Centrifuge tubes, Oak Ridge, Nalgene®
Supplier: Thermo Fisher Scientific
Serological pipettes, Stripette™
Supplier: Corning
Multiple well cell culture plates, Costar®
Supplier: Corning
Cell culture flasks with standard or filter caps, CELLSTAR®
Supplier: Greiner Bio-One
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Nalgene®, Biotainer® Bottles
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Heat sealing foil, ThermASeal Foil™
Supplier: iST Scientific Ltd
System Boxes
Supplier: KOEHLER TECHNISCHE PRODUKTEN
CELLSTAR PLATE CELL REPELLENT 96W 39 1 * 32 items
Supplier: Greiner Bio-One