AMPIVIEW HYBRIDIZATION BUFFER (1X) 1 * 10 mL
Catalog # ENZOENZACC1520010
Supplier: ENZO LIFE SCIENCES
AMPIVIEW HYBRIDIZATION BUFFER (1X) 1 * 10 mL
Catalog # ENZOENZACC1520010
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
MicroWell™ Plates, 96 well, Nunc™
Supplier: Thermo Fisher Scientific
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
CostarRegistered Mark 12 mm TranswellRegistered Mark, 0.4 µm Pore Polyester Membrane Inserts 1 * 48 items
Supplier: STEMCELL Technologies
VWR® Safe-T-Flex, Caps for Test Tubes
Supplier: VWR Collection
BOTTLE PET 1L SQ STER 18MG WHITE DOME CAP 1 * 84 items
Supplier: LAB3 OWN PRODUCTION
VWR®, Cell Culture Insert Plates, PET
Supplier: VWR Collection
AriaMx real-time PCR system
Supplier: AGILENT
Mass cell culture system, CELLdisc™
Supplier: Greiner Bio-One
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Supplier: Simport Scientific