OXYNTOMODULIN MONOCLONAL ANTIBODY (46) 1 * 1 mg
Catalog # ENZOBPDABS064461
Supplier: ENZO LIFE SCIENCES
OXYNTOMODULIN MONOCLONAL ANTIBODY (46) 1 * 1 mg
Catalog # ENZOBPDABS064461
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR® Drosophila Vials
Supplier: VWR Collection
Multiple well cell culture plates, Costar®
Supplier: Corning
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Inoculating loops and needles, disposable
Supplier: Greiner Bio-One
FalconRegistered Mark Round-Bottom Tubes, 14 mL 1 * 500 items
Supplier: STEMCELL Technologies
Scoops, long handled, SteriWare®
Supplier: Sampling Systems
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Transfer pipettes, narrow stem, Samco™
Supplier: SAMCO
Cell culture flask with quick release cap
Supplier: SARSTEDT
Measuring scoops
Supplier: VITLAB
PCR tubes, 0,1 ml, with or without cap strips, Axygen®
Supplier: Corning
Bioassay dish, Nunclon™Δ
Supplier: Thermo Fisher Scientific
1536-Well cyclic olefin copolymer microplates, Corning®
Supplier: Corning
384-well (HTS) microplates, non treated, BRANDplates®
Supplier: Brand