TENASCIN (HUMAN) MONOCLONAL ANTIBODY (DB7) 1 * 100 µl
Catalog # ENZOBMLTG60440100
Supplier: ENZO LIFE SCIENCES
TENASCIN (HUMAN) MONOCLONAL ANTIBODY (DB7) 1 * 100 µl
Catalog # ENZOBMLTG60440100
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
VWR®, Aluminium Foil Seals for Cold Storage
Supplier: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Half Size, 136×66,5 mm
Supplier: VWR Collection
VWR® Electroporation Cuvettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR® Water and General Sampling Bottles
Supplier: VWR Collection
VWR®, Stoppers, PE
Supplier: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Conical Sample Containers, with Snap-On Lids
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Spreaders, L-Shaped, Standard Line
Supplier: VWR Collection
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
VWR®, Cryo Boxes
Supplier: VWR Collection
Containers with snap caps, Corning® Gosselin™
Supplier: Corning
Glass bottom microplates, SensoPlate™
Supplier: Greiner Bio-One
VWR®, Serological Pipettes, Open End
Supplier: VWR Collection