KANAMYCIN SOLUTION (100MG/ML) 1 * 100 mL
Catalog # APOSBIG1036-100ML
Supplier: Apollo Scientific
Frequently Bought Together
Sampling scoops, SteriPlast®
Supplier: BURKLE
VWR®, Petri Dish, PS
Supplier: VWR Collection
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Sealing tape for MicroWell™ plates, Nunc™
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
VWR®, Centrifuge Tubes, Metal-Free, Sterile
Supplier: VWR Collection
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Chloroform 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific