Order Entry
Ireland
ContactUsLinkComponent
C25Z-2144EUHB - 4 mpos dead-end path, u-electric, USB 1/16*.4mm, 50C/250psi liq, PEEK E4/E2 Universal electric actuator Serial interface: USB 1 * 1 items
Catalog # VICIC25Z-2144EUHB
Supplier:  VICI
C25Z-2144EUHB - 4 mpos dead-end path, u-electric, USB 1/16*.4mm, 50C/250psi liq, PEEK E4/E2 Universal electric actuator Serial interface: USB 1 * 1 items
Catalog # VICIC25Z-2144EUHB
Supplier:  VICI
Frequently Bought Together