Order Entry
Ireland
ContactUsLinkComponent
Support frame, for 1100 mm wide under bench cabinet, sheet steel powder-coated/melamin resin laminated, RAL 7035 light grey for hazardous materials work stations with width 1200 mm with depth 600 mm, for model(s): GAP Version: SV version - standing w 1 * 1 items
Catalog # MSGA41076
Supplier:  asecos
Support frame, for 1100 mm wide under bench cabinet, sheet steel powder-coated/melamin resin laminated, RAL 7035 light grey for hazardous materials work stations with width 1200 mm with depth 600 mm, for model(s): GAP Version: SV version - standing w 1 * 1 items
Catalog # MSGA41076
Supplier:  asecos
Frequently Bought Together