Order Entry
Ireland
ContactUsLinkComponent
1200 Infinity Series Instant Pilot Handheld local controller for all Agilent HPLC systems with color screen, keyboard, and holder. Includes CAN cable and USB slot for method and sequence transfer. 1 * 1 items
Catalog # HEWLG4208A
Supplier:  Agilent
1200 Infinity Series Instant Pilot Handheld local controller for all Agilent HPLC systems with color screen, keyboard, and holder. Includes CAN cable and USB slot for method and sequence transfer. 1 * 1 items
Catalog # HEWLG4208A
Supplier:  Agilent
Frequently Bought Together