0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
CHLORTALIDONE FOR SYSTEM SUITABILITY 1 * 1,2 mg
EDQMY0002129
:
EDQM
CHLORTALIDONE FOR SYSTEM SUITABILITY 1 * 1,2 mg
EDQMY0002129
:
EDQM
Frequently Bought Together
SpecCheck secondary gel standards sets
: Hach
Supelco® PESTANAL® Organic Reference Standard, Malaoxon
: Merck
Rotigotine Related Compounds, Reference Standard
: USP
pH buffers, for LAQUAtwin meters
: HORIBA
VWR®, Sulphur, Certified Reference Materials, in heavy mineral oil
: VWR Chemicals
Pharmaceutical secondary standard, telmisartan, (CRM), Supelco®
: Merck
VWR®, High temperature viscosity standards
: VWR Chemicals
IODOMETHANE 1 * 1 mL
: Merck
Supelco® PESTANAL® Organic Reference Standard, Phenmedipham
: Merck
Nickel standard solution, 1000 mg/L Ni in dil. nitric acid standard for ICP
: CPACHEM
2-Butanol, Reference Standard
: USP