KLERCIDE SPOR.LOW RES.PEROXIDE 5L STER 1 * 4 items
Catalog # SHIE6600548
Supplier: ECOLAB CONTAMINATION CONTROL
KLERCIDE SPOR.LOW RES.PEROXIDE 5L STER 1 * 4 items
Catalog # SHIE6600548
Supplier: ECOLAB CONTAMINATION CONTROL
Frequently Bought Together
Acrylonitrile 99+%
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Supplier: Honeywell Chemicals
ISE standards
Supplier: Hach
1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Butyl acetate 99+%, extra pure
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclohexane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
tert-Butanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals