Vesta-Syde SQ Sterile Quat Disinfectant, 3.78L x 4 1 * 4 items
Catalog # STER634008EC
Supplier: STERIS
Vesta-Syde SQ Sterile Quat Disinfectant, 3.78L x 4 1 * 4 items
Catalog # STER634008EC
Supplier: STERIS
Frequently Bought Together
Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
Supplier: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
N,N-Dimethylformamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene ≥99%, GPR RECTAPUR® for pathology
Supplier: VWR Chemicals
Ethanol 20% (w/w), Emprove® Expert for cleaning biochromatography resins, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Acetone, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals