Order Entry
Ireland
ContactUsLinkComponent
The Quenching Solution is intended to be used to reduce autofluorescence in FISH on formalin-fixed, paraffin-embedded specimens. 1 * 8 mL
Catalog # ABNOU0530.8ML
Supplier:  Abnova
The Quenching Solution is intended to be used to reduce autofluorescence in FISH on formalin-fixed, paraffin-embedded specimens. 1 * 8 mL
Catalog # ABNOU0530.8ML
Supplier:  Abnova
Frequently Bought Together