Order Entry
Ireland
ContactUsLinkComponent
Yersinia IgA Rapid Test is for qualitive and semi-quantitative of Yersinia spp infection using IgA antibodies in human serum or plasma. 1 * 1 KIT
Catalog # ABNOKA6668.1KIT
Supplier:  Abnova
Yersinia IgA Rapid Test is for qualitive and semi-quantitative of Yersinia spp infection using IgA antibodies in human serum or plasma. 1 * 1 KIT
Catalog # ABNOKA6668.1KIT
Supplier:  Abnova
Frequently Bought Together