Acetonitrile gradient grade for liquid chromatography LiChrosolv Reag. Ph Eur 1 * 185 L
1.00030.9189
: Merck
Acetonitrile gradient grade for liquid chromatography LiChrosolv Reag. Ph Eur 1 * 185 L
1.00030.9189
: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals