OXALATE PH STANDARD SOLUTION 1 * 200 mL
Catalog # ACSDJPH057
Supplier: CPACHEM
Frequently Bought Together
Potassium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Ammonia solution 25%, EMPROVE® ESSENTIAL Ph. Eur., BP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ammonia solution 25%, LiChropur™ for HPLC, Supelco®
Supplier: Merck
Soda lime, AnalaR NORMAPUR® analytical reagent, with indicator
Supplier: VWR Chemicals
VWR® Kjeldahl Tablets
Supplier: VWR Chemicals
pH indicator strips, Whatman™
Supplier: Whatman products (Cytiva)
Nitrite VHR L, 500 ml
Supplier: Lovibond
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach