Terminal disinfectant for open plant application, QAC based 1 * 1 items
Catalog # DIVY101108285
Supplier: Diversey
Terminal disinfectant for open plant application, QAC based 1 * 1 items
Catalog # DIVY101108285
Supplier: Diversey
Frequently Bought Together
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Glycerine 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
2-Butoxyethanol ≥99%
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
1,2-Dichloroethane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific