FILLING SOLUTION 3M KCL (100ML) 1 * 100 mL
Catalog # JENW99959-51
Supplier: Cole-Parmer
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
Supplier: HAMILTON BONADUZ
Sucrose, Reference Standard
Supplier: USP
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Potassium iodide 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1070502
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, TOC standard solutions
Supplier: VWR Chemicals
Turbidity standards, Stablcal®
Supplier: Hach
Supelco® Organic Reference Standard, 1H-1,2,4-Triazole
Supplier: Merck
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Organic reference standard, Trolamine, Ph.Eur., RS
Supplier: EDQM