Order Entry
Ireland
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D & Activated Ionizer and D raft Shield Motors 1 * 1 items
Catalog # SARBMCA66P3CF2H
Supplier:  Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D & Activated Ionizer and D raft Shield Motors 1 * 1 items
Catalog # SARBMCA66P3CF2H
Supplier:  Sartorius Balances
Frequently Bought Together