Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36P3CE24I
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36P3CE24I
Supplier: Sartorius Balances
Frequently Bought Together
Accessories for vortex mixers, MS 3
Supplier: IKA
Distance sieve ZM 300 titanium trapezoid holes 0.25 mm 1 * 1 items
Supplier: Retsch GmbH
Accessories for mortar grinder, RM 200
Supplier: Retsch GmbH
Accessories for Q500/Q700 Sonicators
Supplier: QSONICA LLC
Water baths, unstirred, SUB Aqua Pro series
Supplier: GRANT INSTRUMENTS
Undergrad analogue magnetic hotplate stirrers, Stuart®
Supplier: Cole Parmer
Single height adjustable vessel clamp for Solaris Shaker 11”x14” Platforms 1 * 1 items
Supplier: Thermo Fisher Scientific
Stirrers, propeller, with four paddles
Supplier: BOCHEM
Accessories for laboratory grinding Mills, POLYMIX® System PX-MFC 90 D
Supplier: KINEMATICA
Distance sieve for ZM 300 stainless steel round holes 6.00 mm 1 * 1 items
Supplier: Retsch GmbH
VWR®, Accessories for EML 450 Digital Plus Test Sieve Shakers
Supplier: VWR Collection