Order Entry
Ireland
ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA116S3CF3
Supplier:  Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA116S3CF3
Supplier:  Sartorius Balances
Frequently Bought Together